Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281969.1 | NP_001268898.1 | 5 - 6 | 645 | 98 | |
| NM_010298.6 | NP_034428.2 | 5 - 6 | 695 | 98 | ||
| GenBank mRNA | AK082578.1 | - | 5 - 6 | 698 | 98 | |
| AK083251.1 | - | 4 - 5 | 549 | 98 | ||
| AM420316.1 | - | 4 - 5 | 528 | 98 | ||
| BC027094.1 | - | 5 - 6 | 649 | 98 | ||
| BC037605.1 | - | 5 - 6 | 663 | 5361798 | 98 | |
| U09399.1 | - | 5 - 6 | 604 | 98 | ||
| X81202.1 | - | 4 - 5 | 487 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glrb
Genomic Map