Assay Details
| Gene Symbol: Glrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281969.1 | NP_001268898.1 | 7 - 8 | 876 | 89 | |
| NM_010298.6 | NP_034428.2 | 7 - 8 | 926 | 89 | ||
| GenBank mRNA | AK082578.1 | - | 7 - 8 | 929 | 89 | |
| AK083251.1 | - | 6 - 7 | 780 | 89 | ||
| BC027094.1 | - | 7 - 8 | 880 | 89 | ||
| BC037605.1 | - | 7 - 8 | 894 | 5361798 | 89 | |
| U09399.1 | - | 7 - 8 | 835 | 89 | ||
| X81202.1 | - | 6 - 7 | 718 | 89 | ||
More Information
Set Membership: |
|

Glrb
Genomic Map