Assay Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170985.1 | NP_001164456.1 | 10 - 11 | 1633 | 77 | |
| NM_130867.3 | NP_570937.2 | 10 - 11 | 1630 | 77 | ||
| GenBank mRNA | AF480411.1 | - | 9 - 10 | 1268 | 77 | |
| AK158381.1 | - | 10 - 11 | 1587 | 77 | ||
| AY243095.1 | - | 9 - 10 | 1268 | 77 | ||
| BC023765.1 | - | 10 - 11 | 1446 | 5343720 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel
Genomic Map