Assay Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170985.1 | NP_001164456.1 | 13 - 14 | 2040 | 69 | |
| NM_130867.3 | NP_570937.2 | 13 - 14 | 2037 | 69 | ||
| GenBank mRNA | AF480411.1 | - | 12 - 13 | 1675 | 69 | |
| AK158381.1 | - | 13 - 14 | 1993 | 69 | ||
| AY243095.1 | - | 12 - 13 | 1675 | 69 | ||
| BC023765.1 | - | 13 - 14 | 1853 | 5343720 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel
Genomic Map