Assay Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170985.1 | NP_001164456.1 | 3 - 4 | 663 | 87 | |
| NM_130867.3 | NP_570937.2 | 3 - 4 | 660 | 87 | ||
| GenBank mRNA | AF480411.1 | - | 2 - 3 | 298 | 87 | |
| AK158381.1 | - | 3 - 4 | 617 | 87 | ||
| AY017368.1 | - | 2 - 3 | 298 | 87 | ||
| AY243095.1 | - | 2 - 3 | 298 | 87 | ||
| BC023765.1 | - | 3 - 4 | 476 | 5343720 | 87 | |
| BY736743.1 | - | 3 - 4 | 446 | 87 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel
Genomic Map