Assay Details
| Gene Symbol: Kirrel | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170985.1 | NP_001164456.1 | 8 - 9 | 1375 | 68 | |
| NM_130867.3 | NP_570937.2 | 8 - 9 | 1372 | 68 | ||
| GenBank mRNA | AF480411.1 | - | 7 - 8 | 1010 | 68 | |
| AK158381.1 | - | 8 - 9 | 1329 | 68 | ||
| AY017368.1 | - | 7 - 8 | 1010 | 68 | ||
| AY243095.1 | - | 7 - 8 | 1010 | 68 | ||
| BC023765.1 | - | 8 - 9 | 1188 | 5343720 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Kirrel
Genomic Map