Assay Details
| Gene Symbol: Kcnab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289450.1 | NP_001276379.1 | 6 - 7 | 414 | 81 | |
| NM_010597.4 | NP_034727.3 | 5 - 6 | 830 | 81 | ||
| GenBank mRNA | AF033003.1 | - | 5 - 6 | 1185 | 81 | |
| AK004666.1 | - | 6 - 7 | 398 | 81 | ||
| AK031362.1 | - | 5 - 6 | 831 | 81 | ||
| AK138467.1 | - | 5 - 6 | 819 | 81 | ||
| AK153200.1 | - | 6 - 7 | 416 | 81 | ||
| BC014701.1 | - | 5 - 6 | 555 | 4482268 | 81 | |
| U65591.1 | - | 5 - 6 | 426 | 81 | ||
| X97281.1 | - | 5 - 6 | 814 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnab1
Genomic Map