Assay Details
| Gene Symbol: Paxbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026110.2 | NP_080386.3 | 3 - 4 | 686 | 77 | |
| GenBank mRNA | AY033907.1 | - | 3 - 4 | 471 | 77 | |
| AY033908.1 | - | 3 - 4 | 471 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Paxbp1
Genomic Map