Assay Details
| Gene Symbol: Agpat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026792.3 | NP_081068.1 | 2 - 3 | 572 | 139 | |
| GenBank mRNA | AK003649.1 | - | 2 - 3 | 316 | 139 | |
| AK082137.1 | - | 2 - 3 | 347 | 139 | ||
| AK089885.1 | - | 2 - 3 | 375 | 139 | ||
| AK152709.1 | - | 2 - 3 | 315 | 139 | ||
| AK152889.1 | - | 2 - 3 | 319 | 139 | ||
| AK155378.1 | - | 3 - 4 | 521 | 139 | ||
| AK170093.1 | - | 2 - 3 | 316 | 139 | ||
| AY161042.1 | - | 2 - 3 | 549 | 139 | ||
| BC031987.1 | - | 2 - 3 | 297 | 3982362 | 139 | |
| BC062903.1 | - | 2 - 3 | 572 | 139 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Agpat5
Genomic Map