Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 11 - 12 | 1225 | 71 | |
| GenBank mRNA | AK035264.1 | - | 11 - 12 | 1158 | 71 | |
| AK149883.1 | - | 11 - 12 | 1229 | 71 | ||
| AK152926.1 | - | 11 - 12 | 1190 | 71 | ||
| AK159649.1 | - | 11 - 12 | 1154 | 71 | ||
| AK160068.1 | - | 11 - 12 | 1148 | 71 | ||
| BC024084.1 | - | 11 - 12 | 1132 | 5101064 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map