Assay Details
| Gene Symbol: Hgsnat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029884.1 | NP_084160.1 | 13 - 14 | 1474 | 155 | |
| GenBank mRNA | AK035264.1 | - | 13 - 14 | 1407 | 155 | |
| AK149883.1 | - | 13 - 14 | 1478 | 155 | ||
| AK152926.1 | - | 13 - 14 | 1439 | 155 | ||
| AK159649.1 | - | 13 - 14 | 1403 | 155 | ||
| AK160068.1 | - | 13 - 14 | 1397 | 155 | ||
| BC024084.1 | - | 13 - 14 | 1381 | 5101064 | 155 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hgsnat
Genomic Map