Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 10 - 11 | 2325 | 141 | |
| GenBank mRNA | AF079557.1 | - | 10 - 11 | 2074 | 141 | |
| AK034542.1 | - | 3 - 4 | 257 | 141 | ||
| AK036656.1 | - | 10 - 11 | 2348 | 141 | ||
| AK088708.1 | - | 7 - 8 | 680 | 141 | ||
| AK164013.1 | - | 10 - 11 | 2312 | 141 | ||
| BC050892.1 | - | 10 - 11 | 2324 | 141 | ||
| BC059827.1 | - | 10 - 11 | 2334 | 141 | ||
| EF382673.1 | - | 8 - 9 | 1034 | 141 | ||
| JQ890227.1 | - | 7 - 8 | 831 | 141 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map