Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 11 - 12 | 2379 | 130 | |
| GenBank mRNA | AF079557.1 | - | 11 - 12 | 2128 | 130 | |
| AK034542.1 | - | 4 - 5 | 311 | 130 | ||
| AK036656.1 | - | 11 - 12 | 2402 | 130 | ||
| AK088708.1 | - | 8 - 9 | 734 | 130 | ||
| AK164013.1 | - | 11 - 12 | 2366 | 130 | ||
| BC050892.1 | - | 11 - 12 | 2378 | 130 | ||
| BC053037.1 | - | 11 - 12 | 2363 | 131 | ||
| BC059827.1 | - | 11 - 12 | 2388 | 130 | ||
| EF382673.1 | - | 9 - 10 | 1088 | 130 | ||
| JQ890227.1 | - | 8 - 9 | 885 | 130 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map