Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 13 - 14 | 2605 | 69 | |
| GenBank mRNA | AF079557.1 | - | 13 - 14 | 2354 | 69 | |
| AK034542.1 | - | 6 - 7 | 537 | 69 | ||
| AK036656.1 | - | 13 - 14 | 2628 | 69 | ||
| AK088708.1 | - | 10 - 11 | 960 | 69 | ||
| AK164013.1 | - | 13 - 14 | 2592 | 69 | ||
| BC050892.1 | - | 13 - 14 | 2604 | 69 | ||
| BC053037.1 | - | 13 - 14 | 2589 | 69 | ||
| BC059827.1 | - | 13 - 14 | 2614 | 69 | ||
| EF382673.1 | - | 11 - 12 | 1314 | 69 | ||
| JQ890227.1 | - | 10 - 11 | 1111 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map