Assay Details
| Gene Symbol: Parg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011960.2 | NP_036090.2 | 14 - 15 | 2692 | 74 | |
| GenBank mRNA | AF079557.1 | - | 14 - 15 | 2441 | 74 | |
| AK034542.1 | - | 7 - 8 | 624 | 74 | ||
| AK036656.1 | - | 14 - 15 | 2715 | 74 | ||
| AK088708.1 | - | 11 - 12 | 1047 | 74 | ||
| AK164013.1 | - | 14 - 15 | 2679 | 74 | ||
| BC050892.1 | - | 14 - 15 | 2691 | 74 | ||
| BC053037.1 | - | 14 - 15 | 2676 | 74 | ||
| BC059827.1 | - | 14 - 15 | 2701 | 74 | ||
| EF382673.1 | - | 12 - 13 | 1401 | 74 | ||
| JQ890227.1 | - | 11 - 12 | 1198 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Parg
Genomic Map