Assay Details
| Gene Symbol: 1700024G13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034037.1 | NP_001029209.1 | 1 - 2 | 116 | 70 | |
| GenBank mRNA | AK006301.1 | - | 1 - 2 | 135 | 70 | |
| AK007014.1 | - | 1 - 2 | 134 | 70 | ||
| AK019799.1 | - | 1 - 2 | 123 | 70 | ||
| BC107222.2 | - | 1 - 2 | 113 | 40051877 | 70 | |
| BC107223.1 | - | 1 - 2 | 116 | 40051878 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

1700024G13Rik
Genomic Map