Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052424.1 | - | 1 - 2 | 225 | 59 | |
| AK164351.1 | - | 1 - 2 | 209 | 59 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052424.1 | - | 1 - 2 | 225 | 59 | |
| AK164351.1 | - | 1 - 2 | 209 | 59 | ||
Set Membership: |
|