Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139145.4 | NP_631884.1 | 1 - 2 | 64 | 66 | |
| GenBank mRNA | AK052424.1 | - | 2 - 3 | 359 | 66 | |
| AK158212.1 | - | 2 - 3 | 181 | 66 | ||
| AK164351.1 | - | 2 - 3 | 343 | 66 | ||
| BC050090.1 | - | 1 - 2 | 65 | 30012868 | 66 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158212.1, BC050090.1, NM_139145.4 transcripts.
Set Membership: |
|

Hlcs
Genomic Map