Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139145.4 | NP_631884.1 | 5 - 6 | 1613 | 60 | |
| GenBank mRNA | AB066227.1 | - | 4 - 5 | 1473 | 60 | |
| AK052424.1 | - | 6 - 7 | 1909 | 60 | ||
| AK158212.1 | - | 6 - 7 | 1730 | 60 | ||
| AK168925.1 | - | 2 - 3 | 508 | 60 | ||
| BC050090.1 | - | 5 - 6 | 1614 | 30012868 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hlcs
Genomic Map