Assay Details
| Gene Symbol: Hlcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_139145.4 | NP_631884.1 | 8 - 9 | 1964 | 154 | |
| GenBank mRNA | AB066227.1 | - | 7 - 8 | 1824 | 154 | |
| AK052424.1 | - | 9 - 10 | 2260 | 154 | ||
| AK158212.1 | - | 9 - 10 | 2081 | 154 | ||
| AK168925.1 | - | 5 - 6 | 859 | 154 | ||
| BC050090.1 | - | 8 - 9 | 1965 | 30012868 | 154 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hlcs
Genomic Map