Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400002305.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002305.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002309.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002309.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002303.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002303.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002307.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002307.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002312.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002312.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002310.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002310.mm.1 |
TC1400000281.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400002301.mm.1 |
NM_177814.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400002301.mm.1 |
TC1400000281.mm.1 |
MTA TC |