Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 10 - 11 | 1425 | 71 | |
| GenBank mRNA | AK087583.1 | - | 2 - 3 | 146 | 71 | |
| AK145297.1 | - | 9 - 10 | 1438 | 71 | ||
| AK158666.1 | - | 8 - 9 | 943 | 71 | ||
| AK170006.1 | - | 8 - 9 | 892 | 71 | ||
| BC070471.1 | - | 10 - 11 | 1425 | 30626499 | 71 | |
| BC072583.1 | - | 2 - 3 | 542 | 6403859 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc66
Genomic Map