Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 11 - 12 | 1585 | 83 | |
| GenBank mRNA | AK087583.1 | - | 3 - 4 | 306 | 83 | |
| AK158666.1 | - | 9 - 10 | 1103 | 83 | ||
| AK170006.1 | - | 9 - 10 | 1052 | 83 | ||
| BC070471.1 | - | 11 - 12 | 1585 | 30626499 | 83 | |
| BC072583.1 | - | 3 - 4 | 702 | 6403859 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc66
Genomic Map