Assay Details
| Gene Symbol: Ccdc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177111.3 | NP_796085.3 | 14 - 15 | 2332 | 92 | |
| GenBank mRNA | AK087583.1 | - | 6 - 7 | 1053 | 92 | |
| AK158666.1 | - | 12 - 13 | 1850 | 92 | ||
| AK170006.1 | - | 12 - 13 | 1799 | 92 | ||
| BC070471.1 | - | 14 - 15 | 2333 | 30626499 | 92 | |
| BC072583.1 | - | 6 - 7 | 1449 | 6403859 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ccdc66
Genomic Map