Assay Details
| Gene Symbol: Arhgef3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289687.1 | NP_001276616.1 | 1 - 2 | 230 | 101 | |
| GenBank mRNA | BC005517.1 | - | 1 - 2 | 201 | 3494309 | 101 |
| BC012262.1 | - | 1 - 2 | 182 | 4192378 | 101 | |
| CD539300.1 | - | 1 - 2 | 220 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Arhgef3
Genomic Map