Assay Details
| Gene Symbol: Obp1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008754.2 | NP_032780.2 | 6 - 7 | 548 | 110 | |
| GenBank mRNA | AK017455.1 | - | 6 - 7 | 549 | 110 | |
| BC125514.1 | - | 6 - 7 | 535 | 40130029 | 110 | |
| BC125516.1 | - | 6 - 7 | 535 | 40130031 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978013 | NM_008754.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1978013 | AK017455.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 120905 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 756145 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 125367 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978013 | NM_008754.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1978013 | AK017455.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P510420 | AK017455.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480293 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480280 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480291 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480281 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480289 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480267 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480277 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480290 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480269 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32480278 | - | - |
| Whole Mouse Genome G4122A | A_51_P510420 | AK017455.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104209 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P09337564 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14714640 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14714650 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14714637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480257 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480251 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480253 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480280 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480244 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480254 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480290 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480243 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480249 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480256 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480259 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480293 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32480319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480260 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480323 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14596248 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480258 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15158980 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480257 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480247 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14596243 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480269 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480291 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480256 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480254 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480326 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14714639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480289 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480267 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480278 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480253 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480290 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14714646 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480319 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480272 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15200679 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P15200680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480282 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32480251 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10605492 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605489 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605490 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10605491 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1453644_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10605488 | - | - |
| Mouse Genome 430 2.0 Array | 1453644_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1453644_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18249_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 18249_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00016020.mm.1 | NM_008754.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016020.mm.1 | NM_008754.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016020.mm.1 | TC0X00002510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00016021.mm.1 | NM_008754.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016021.mm.1 | NM_008754.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016021.mm.1 | TC0X00002510.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00016018.mm.1 | NM_008754.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016018.mm.1 | NM_008754.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00016018.mm.1 | TC0X00002510.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1453644_PM_at | - | - |
| Mouse Expression Set 430 | 1453644_at | - | - |
| Murine Genome U74v2 Set | 99377_PM_at | - | - |
| Murine Genome U74v2 Set | 99377_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453644_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1453644_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453644_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1453644_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 872188 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 5860280 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005860280 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2694682 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2694682 | - | - |
| mouse-6 v1.1 (Illumina) | scl54102.7.1_33-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5860280 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54102.7.1_33-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5860280 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2694682 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2694682 | - | - |

Obp1a
Genomic Map