Assay Details
| Gene Symbol: Rbks | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153196.1 | NP_694876.1 | 4 - 5 | 361 | 66 | |
| GenBank mRNA | AK017229.1 | - | 4 - 5 | 379 | 66 | |
| AK138939.1 | - | 4 - 5 | 379 | 66 | ||
| BC023339.1 | - | 4 - 5 | 361 | 5253827 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rbks
Genomic Map