Assay Details
| Gene Symbol: Ckap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004140.2 | NP_001004140.2 | 3 - 4 | 283 | 162 | |
| GenBank mRNA | AK132463.1 | - | 3 - 4 | 271 | 162 | |
| AY692438.1 | - | 3 - 4 | 283 | 162 | ||
| BC145666.1 | - | 3 - 4 | 276 | 40130969 | 162 | |
| BC146023.1 | - | 3 - 4 | 276 | 40131326 | 162 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ckap2
Genomic Map