Assay Details
| Gene Symbol: Ppp2r2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172994.2 | NP_766582.1 | 6 - 7 | 1037 | 55 | |
| GenBank mRNA | AK032023.1 | - | 6 - 7 | 940 | 55 | |
| AK049830.1 | - | 6 - 7 | 923 | 55 | ||
| AY348866.1 | - | 6 - 7 | 1039 | 55 | ||
| BC059811.1 | - | 6 - 7 | 1037 | 6849535 | 55 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r2c
Genomic Map