Assay Details
| Gene Symbol: Sis | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081137.1 | NP_001074606.1 | 31 - 32 | 3861 | 110 | |
| GenBank mRNA | EU937530.1 | - | 32 - 33 | 3924 | 110 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sis
Genomic Map