Assay Details
| Gene Symbol: Ift80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026641.2 | NP_080917.1 | 15 - 16 | 1843 | 125 | |
| GenBank mRNA | AK019542.1 | - | 2 - 3 | 1760 | 125 | |
| AK042231.1 | - | 15 - 16 | 1768 | 125 | ||
| AK147305.1 | - | 14 - 15 | 5370 | 125 | ||
| BC013814.1 | - | 10 - 11 | 1132 | 125 | ||
| BC034101.1 | - | 15 - 16 | 1738 | 4501451 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ift80
Genomic Map