Assay Details
| Gene Symbol: Smc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133786.3 | NP_598547.1 | 12 - 13 | 1975 | 88 | |
| GenBank mRNA | AJ534940.2 | - | 11 - 12 | 1851 | 88 | |
| AK048650.1 | - | 7 - 8 | 1188 | 88 | ||
| BC061481.1 | - | 12 - 13 | 1950 | 6512832 | 88 | |
| BC062939.1 | - | 12 - 13 | 1975 | 6841276 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Smc4
Genomic Map