Assay Details
| Gene Symbol: Appl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145221.2 | NP_660256.1 | 6 - 7 | 514 | 130 | |
| GenBank mRNA | AK030445.1 | - | 6 - 7 | 394 | 130 | |
| AK033566.1 | - | 6 - 7 | 485 | 130 | ||
| AK045438.1 | - | 6 - 7 | 450 | 130 | ||
| AK049307.1 | - | 6 - 7 | 501 | 130 | ||
| AK088080.1 | - | 6 - 7 | 465 | 130 | ||
| AK146356.1 | - | 6 - 7 | 485 | 130 | ||
| AY113705.1 | - | 6 - 7 | 479 | 130 | ||
| BC063751.1 | - | 6 - 7 | 526 | 6511401 | 130 | |
| BU514763.1 | - | 6 - 7 | 525 | 130 | ||
More Information
Set Membership: |
|

Appl1
Genomic Map