Assay Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178880.4 | NP_849211.3 | 4 - 5 | 6452 | 158 | |
| GenBank mRNA | AB546195.1 | - | 5 - 6 | 6267 | 158 | |
| AK122418.1 | - | 2 - 3 | 813 | 158 | ||
| BC145121.1 | - | 4 - 5 | 6394 | 158 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Son
Genomic Map