Assay Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178880.4 | NP_849211.3 | 5 - 6 | 6611 | 72 | |
| GenBank mRNA | AB546195.1 | - | 6 - 7 | 6426 | 72 | |
| AK122418.1 | - | 3 - 4 | 972 | 72 | ||
| BC145121.1 | - | 5 - 6 | 6553 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Son
Genomic Map