Assay Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178880.4 | NP_849211.3 | 6 - 7 | 6801 | 112 | |
| GenBank mRNA | AK122418.1 | - | 4 - 5 | 1162 | 112 | |
| BC145121.1 | - | 6 - 7 | 6743 | 112 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Son
Genomic Map