Assay Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178880.4 | NP_849211.3 | 8 - 9 | 7028 | 68 | |
| GenBank mRNA | BC145121.1 | - | 8 - 9 | 6970 | 68 | |
| CB849818.2 | - | 1 - 2 | 126 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Son
Genomic Map