Assay Details
| Gene Symbol: Ifngr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008338.3 | NP_032364.1 | 5 - 6 | 777 | 72 | |
| GenBank mRNA | AK137165.1 | - | 5 - 6 | 800 | 72 | |
| AK143214.1 | - | 5 - 6 | 792 | 72 | ||
| AK146995.1 | - | 795 | 72 | |||
| AK165193.1 | - | 5 - 6 | 793 | 72 | ||
| AK169907.1 | - | 5 - 6 | 798 | 72 | ||
| BC055745.1 | - | 5 - 6 | 777 | 6403454 | 72 | |
| S69336.1 | - | 5 - 6 | 801 | 72 | ||
More Information
Set Membership: |
|

Ifngr2
Genomic Map