Assay Details
| Gene Symbol: Mmp16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019724.3 | NP_062698.2 | 4 - 5 | 949 | 68 | |
| GenBank mRNA | AB021228.1 | - | 4 - 5 | 764 | 68 | |
| AF282844.1 | - | 4 - 5 | 716 | 68 | ||
| AK046170.1 | - | 4 - 5 | 808 | 68 | ||
| AK048543.1 | - | 4 - 5 | 829 | 68 | ||
| AK053598.1 | - | 4 - 5 | 839 | 68 | ||
| AK077939.1 | - | 992 | 68 | |||
| BC057926.1 | - | 4 - 5 | 951 | 5292478 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mmp16
Genomic Map