Assay Details
| Gene Symbol: Cngb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013927.2 | NP_038955.1 | 14 - 15 | 1721 | 105 | |
| GenBank mRNA | AJ243572.1 | - | 14 - 15 | 1721 | 105 | |
| AK040140.1 | - | 5 - 6 | 588 | 105 | ||
More Information
Set Membership: |
|

Cngb3
Genomic Map