| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400018846.mm.1 |
NM_001276292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018846.mm.1 |
NM_177327.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018846.mm.1 |
NR_074089.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018846.mm.1 |
NR_074090.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018846.mm.1 |
TC0400002271.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400018847.mm.1 |
NM_001276292.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018847.mm.1 |
NM_177327.5 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018847.mm.1 |
NR_074089.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018847.mm.1 |
NR_074090.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400018847.mm.1 |
TC0400002271.mm.1 |
MTA TC |