Assay Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276292.1 | NP_001263221.1 | 5 - 6 | 531 | 82 | |
| NM_177327.5 | NP_796301.2 | 4 - 5 | 488 | 82 | ||
| NR_074089.1 | 4 - 5 | 488 | 82 | |||
| GenBank mRNA | AK032232.1 | - | 4 - 5 | 479 | 82 | |
| AK042432.1 | - | 4 - 5 | 479 | 82 | ||
| AK137588.1 | - | 4 - 5 | 413 | 82 | ||
| BB662323.1 | - | 4 - 5 | 482 | 82 | ||
| BC026829.1 | - | 5 - 6 | 469 | 4209397 | 82 | |
| BC150812.1 | - | 3 - 4 | 323 | 9087723 | 82 | |
| CJ137158.1 | - | 4 - 5 | 448 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC026829.1, NM_001276292.1 transcripts.
Set Membership: |
|

Wwp1
Genomic Map