Assay Details
| Gene Symbol: Wwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276292.1 | NP_001263221.1 | 8 - 9 | 1166 | 117 | |
| NM_177327.5 | NP_796301.2 | 9 - 10 | 1328 | 117 | ||
| NR_074090.1 | 9 - 10 | 1312 | 117 | |||
| GenBank mRNA | AK033138.1 | - | 1 - 2 | 142 | 117 | |
| AK137588.1 | - | 9 - 10 | 1253 | 117 | ||
| AK168567.1 | - | 9 - 10 | 1279 | 117 | ||
| AK191481.1 | - | 1 - 2 | 40 | 117 | ||
| BC026829.1 | - | 8 - 9 | 1104 | 4209397 | 117 | |
| BC150812.1 | - | 8 - 9 | 1163 | 9087723 | 117 | |
More Information
Set Membership: |
|

Wwp1
Genomic Map