Assay Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133907.3 | NP_598668.1 | 17 - 18 | 2503 | 126 | |
| GenBank mRNA | AK029973.1 | - | 17 - 18 | 2313 | 126 | |
| AK046056.1 | - | 17 - 18 | 2504 | 126 | ||
| AK122187.1 | - | 17 - 18 | 2438 | 126 | ||
| BC021525.1 | - | 6 - 7 | 748 | 126 | ||
| BC059245.1 | - | 17 - 18 | 2318 | 126 | ||
| BC120731.1 | - | 17 - 18 | 2250 | 40129654 | 126 | |
| BC137626.1 | - | 17 - 18 | 2250 | 8860645 | 126 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube3c
Genomic Map