Assay Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133907.3 | NP_598668.1 | 18 - 19 | 2750 | 75 | |
| GenBank mRNA | AK029973.1 | - | 18 - 19 | 2560 | 75 | |
| AK046056.1 | - | 18 - 19 | 2751 | 75 | ||
| AK049125.1 | - | 1 - 2 | 141 | 75 | ||
| AK122187.1 | - | 18 - 19 | 2685 | 75 | ||
| AK203997.1 | - | 1 - 2 | 221 | 75 | ||
| BC021525.1 | - | 7 - 8 | 995 | 75 | ||
| BC059245.1 | - | 18 - 19 | 2565 | 75 | ||
| BC120731.1 | - | 18 - 19 | 2497 | 40129654 | 75 | |
| BC137626.1 | - | 18 - 19 | 2497 | 8860645 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube3c
Genomic Map