Assay Details
| Gene Symbol: Ube3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133907.3 | NP_598668.1 | 20 - 21 | 3154 | 68 | |
| GenBank mRNA | AK029973.1 | - | 20 - 21 | 2964 | 68 | |
| AK046056.1 | - | 20 - 21 | 3155 | 68 | ||
| AK049125.1 | - | 3 - 4 | 545 | 68 | ||
| AK122187.1 | - | 20 - 21 | 3089 | 68 | ||
| BC021525.1 | - | 9 - 10 | 1399 | 68 | ||
| BC059245.1 | - | 20 - 21 | 2969 | 68 | ||
| BC120731.1 | - | 20 - 21 | 2901 | 40129654 | 68 | |
| BC137626.1 | - | 20 - 21 | 2901 | 8860645 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube3c
Genomic Map