| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0500033280.mm.1 |
TC0500003626.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033280.mm.1 |
NM_010229.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033280.mm.1 |
NM_010229.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033282.mm.1 |
TC0500003626.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033282.mm.1 |
NM_010229.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033282.mm.1 |
NM_010229.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033281.mm.1 |
TC0500003626.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0500033281.mm.1 |
NM_010229.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0500033281.mm.1 |
NM_010229.2 |
Refseq |