Assay Details
| Gene Symbol: Map3k7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009316.1 | NP_033342.1 | 8 - 9 | 1022 | 116 | |
| NM_172688.3 | NP_766276.1 | 8 - 9 | 1022 | 116 | ||
| GenBank mRNA | AK046565.1 | - | 8 - 9 | 988 | 116 | |
| AK053911.1 | - | 8 - 9 | 1008 | 116 | ||
| AK133642.1 | - | 2 - 3 | 149 | 116 | ||
| AK137046.1 | - | 8 - 9 | 1023 | 116 | ||
| AK159277.1 | - | 8 - 9 | 986 | 116 | ||
| BC006665.1 | - | 8 - 9 | 1013 | 3499247 | 116 | |
| CV559615.1 | - | 1 - 2 | 78 | 116 | ||
| D76446.1 | - | 8 - 9 | 1022 | 116 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Map3k7
Genomic Map