Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293703.1 | NP_001280632.1 | 2 - 3 | 216 | 72 | |
| NM_028000.1 | NP_082276.1 | 2 - 3 | 216 | 72 | ||
| GenBank mRNA | AA184457.1 | - | 2 - 3 | 216 | 72 | |
| AK009455.1 | - | 2 - 3 | 183 | 72 | ||
| AK144593.1 | - | 2 - 3 | 201 | 72 | ||
| BB857717.1 | - | 2 - 3 | 217 | 72 | ||
| BC099489.1 | - | 2 - 3 | 167 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppapdc1b
Genomic Map