Assay Details
| Gene Symbol: Ppapdc1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293703.1 | NP_001280632.1 | 4 - 5 | 368 | 82 | |
| NM_028000.1 | NP_082276.1 | 4 - 5 | 368 | 82 | ||
| NR_121642.1 | 4 - 5 | 328 | 82 | |||
| NR_121643.1 | 4 - 5 | 328 | 82 | |||
| GenBank mRNA | AA184457.1 | - | 4 - 5 | 368 | 82 | |
| AK007542.1 | - | 2 - 3 | 329 | 82 | ||
| AK009455.1 | - | 4 - 5 | 335 | 82 | ||
| AK144593.1 | - | 4 - 5 | 353 | 82 | ||
| BB857717.1 | - | 4 - 5 | 369 | 82 | ||
| BC023111.1 | - | 4 - 5 | 315 | 82 | ||
| BC039626.1 | - | 3 - 4 | 183 | 82 | ||
| BC062173.1 | - | 4 - 5 | 301 | 82 | ||
| BC089556.1 | - | 4 - 5 | 312 | 82 | ||
| BC099489.1 | - | 4 - 5 | 319 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppapdc1b
Genomic Map